missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Mixed lineage kinase domain-like Protein
A DNA sequence encoding the Mus musculus (Mouse) Mixed lineage kinase domain-like protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9886-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001297542.1 | |
Mixed lineage kinase domain-like Protein | |
Research Use Only | |
MLKL | |
Recombinant Protein | |
Yeast | |
MDKLGQIIKL GQLIYEQCEK MKYCRKQCQR LGNRVHGLLQ PLQRLQAQGK KNLPDDITAA LGRFDEVLKE ANQQIEKFSK KSHIWKFVSV GNDKILFHEV NEKLRDVWEE LLLLLQVYHW NTVSDVSQPA SWQQEDRQDA EEDGNENMKV ILMQLQISVE EINKTLKQCS LKPTQEIPQD LQIKEIPKEH LGPPWTKLKT SKMSTIYRGE YHRSPVTIKV FNNPQAESVG IVRFTFNDEI KTMKKFDSPN ILRIFGICID QTVKPPEFSI VMEYCELGTL RELLDREKDL TMSVRSLLVL RAARGLYRLH HSETLHRNIS SSSFLVAGGY QVKLAGFELS KTQNSISRTA KSTKAERSSS TIYVSPERLK NPFCLYDIKA EIYSFGIVLW EIATGKIPFE GCDSKKIREL VAEDKKQEPV GQDCPELLRE IINECRAHEP SQRPSVDGRS LSGRERILER LSAVEESTDK KV | |
Greater than 90% as determined by SDS-PAGE. |
74568 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |