missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Mixed lineage kinase domain-like Protein

A DNA sequence encoding the Mus musculus (Mouse) Mixed lineage kinase domain-like protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9886-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16018921

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001297542.1
Mixed lineage kinase domain-like Protein
Research Use Only
MLKL
Recombinant Protein
Yeast
MDKLGQIIKL GQLIYEQCEK MKYCRKQCQR LGNRVHGLLQ PLQRLQAQGK KNLPDDITAA LGRFDEVLKE ANQQIEKFSK KSHIWKFVSV GNDKILFHEV NEKLRDVWEE LLLLLQVYHW NTVSDVSQPA SWQQEDRQDA EEDGNENMKV ILMQLQISVE EINKTLKQCS LKPTQEIPQD LQIKEIPKEH LGPPWTKLKT SKMSTIYRGE YHRSPVTIKV FNNPQAESVG IVRFTFNDEI KTMKKFDSPN ILRIFGICID QTVKPPEFSI VMEYCELGTL RELLDREKDL TMSVRSLLVL RAARGLYRLH HSETLHRNIS SSSFLVAGGY QVKLAGFELS KTQNSISRTA KSTKAERSSS TIYVSPERLK NPFCLYDIKA EIYSFGIVLW EIATGKIPFE GCDSKKIREL VAEDKKQEPV GQDCPELLRE IINECRAHEP SQRPSVDGRS LSGRERILER LSAVEESTDK KV
Greater than 90% as determined by SDS-PAGE.
74568
200 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers