missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Nmes1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Nmes1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7786-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030791

Quantity Price
1 € 352.00 / 50µg
6 € 328.49 / 50µg
Estimated Shipment
03-11-2020
Add to basket


Specifications

Specifications

NP_001004174.1
Nmes1 Protein
50 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
433470
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Nmes1
Recombinant Protein
E. coli
MGVFQILMKN KELIPLAFFI SVAATGATSF ALYALKKTDV VIDRKRNPEP WEMVDPTQPQ KLITINQQWK PVEELQKVRR ATR
Videos
SDS
Documents