missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nmes1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nmes1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7786-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001004174.1 | |
Nmes1 Protein | |
Research Use Only | |
Nmes1 | |
Recombinant Protein | |
E. coli | |
MGVFQILMKN KELIPLAFFI SVAATGATSF ALYALKKTDV VIDRKRNPEP WEMVDPTQPQ KLITINQQWK PVEELQKVRR ATR | |
Greater than 90% as determined by SDS-PAGE. |
433470 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |