missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse Noggin Protein

Product Code. 18454592 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
This item is not returnable. View return policy

Product Code. 18454592

Brand: Novus Biologicals™ NBP235098500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Noggin protein is fully biologically active when compared to standard. The ED50 as determined by inhibiting BMP-4-induced alkaline phosphatase production of murine ATDC5 cells is less than 2 ng/mL, corresponding to a specific activity of >5.0 x 10^5 IU/mg in the presence of 5 ng/mL BMP-4
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P97466, P97466, P97466
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 30 % acetonitrile, 0.1 % TFA.
Gene ID (Entrez) 9241
Molecular Weight (g/mol) 46.4 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of Noggin as determined by LAL method.
Gene Alias noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1
Gene Symbol NOG
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in 10 mM HAc to a concentration less than 0.25mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Biologically Active Proteins, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein Noggin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.