missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NOV Protein
A cDNA sequence encoding the NOV was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10796-1mg
Additional Details : Weight : 0.01000kg
Specifications
NOV Protein | |
Research Use Only | |
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 1.0 ?g/ml, corresponding to a specific activity of > 1000 IU/mg. | |
Mouse | |
Untagged | |
NOV protein was lyophilized from a 0.2microm filtered concentrated solution in 20mM Tris-HCl, pH 8.5 and 150mM NaCl. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
QVSASLRCPS RCPPKCPSIS PTCAPGVRSV LDGCSCCPVC ARQRGESCSE MRPCDQSSGL YCDRSADPNN QTGICMVPEG DNCVFDGVIY RNGEKFEPNC QYFCTCRDGQ IGCLPRCQLD VLLPGPDCPA PRKVAVPGEC CEKWTCGSDE QGTQGTLGGL ALPAYRPEAT VGVEVSDSSI NCIEQTTEWS ACSKSCGMGV STRVTNRNRQ CEMVKQTRLC IVRPCEQEPE EVTDKKGKKC LRTKKSLKAI HLQFENCTSL YTYKPRFCGV CSDGRCCTPH NTKTIQVEFQ CLPGEIIKKP VMVIGTCTCY SNCPQNNEAF LQDLELKTSR GEI | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |