missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NT5E (CD73) Protein
A cDNA sequence encoding the NT5E (CD73) was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10799-5ug
Additional Details : Weight : 0.01000kg
Specifications
NT5E (CD73) Protein | |
Research Use Only | |
Specific activity is > 8,000 pmol/min/ug, and is defined as the amount of enzyme that hydrolyze 1.0 pmole of Adenosine 5-monophosphate to phosphate per minute per minute at pH 7.5 at 25C. | |
Mouse | |
His | |
NT5E protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
WELTILHTND VHSRLEQTSD DSTKCLNASL CVGGVARLFT KVQQIRKEEP NVLFLDAGDQ YQGTIWFTVY KGLEVAHFMN ILGYDAMALG NHEFDNGVEG LIDPLLRNVK FPILSANIKA RGPLAHQISG LFLPSKVLSV GGEVVGIVGY TSKETPFLSN PGTNLVFEDE ISALQPEVDK LKTLNVNKII ALGHSGFEMD KLIAQKVRGV DIVVGGHSNT FLYTGNPPSK EVPAGKYPFI VTADDGRQVP VVQAYAFGKY LGYLKVEFDD KGNVITSYGN PILLNSSIPE DATIKADINQ WRIKLDNYST QELGRTIVYL DGSTQTCRFR ECNMGNLICD AMINNNLRHP DEMFWNHVSM CIVNGGGIRS PIDEKNNGTI TWENLAAVLP FGGTFDLVQL KGSTLKKAFE HSVHRYGQST GEFLQVGGIH VVYDINRKPW NRVVQLEVLC TKCRVPIYEP LEMDKVYKVT LPSYLANGGD GFQMIKDELL KHDSGDQDIS VVSEYISKMK VVYPAVEGRI KFSLEHHHHH H | |
Greater than 90% as determined by SDS-PAGE. |