missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse OLT-2/TARM1 Fc Chimera Protein

Product Code. 18749306
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18749306

Brand: R&D Systems™ 10000TA050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Recombinant Mouse OLT-2/TARM1 Fc Chimera Protein is derived from NS0

  • TARM-1 (T cell interacting, activating receptor on myeloid cells-1), also name OLT-2 (OSCAR-like Transcript 2), is a type 1 membrane protein expressed by neutrophils, inflammatory monocytes, macrophages, and dendritic cells.
  • Mature mouse TARM-1 contains a 242 amino acid (aa) long extracellular domain with two Ig-like C2 domains, a 21 aa long transmembrane domain and a 9 aa long cytoplasmic domain.
  • The extracellular domain of mouse TARM-1 shares 81.6% and 46.5% aa identity with rat and human TARM-1 respectively.
  • TARM-1 associates with the ITAM bearing adaptor FcR gamma, but not with DAP10 or DAP12.
  • TARM1 expression is also up-regulated by bone marrow-derived macrophages and dendritic cells following stimulation with TLR agonists in vitro.
  • TARM1 receptor stimulation on macrophages and neutrophils co-stimulate the secretion of proinflammatory cytokines induced by TLR ligands, such as LPS.
  • TARM1 Fc fusion protein inhibits anti-CD3 induced CD4 + cell activation, suggesting that TARM1 ectodomain interacts with an unidentified receptor on T cells to inhibit T cell activation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number B6A8R8
For Use With (Application) Bioactivity
Gene ID (Entrez) 441864
Molecular Weight (g/mol) 53 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Mouse myeloma cell line, NS0-derived mouse OLT-2/TARM1 protein

Mouse TARM-1 (Gln17-Ile257) Accession # B6A8R8IEGRMDPMouse IgG2a (Glu98-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.