missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Oxysterol-binding protein-related protein 11 Protein
A DNA sequence encoding the Mus musculus (Mouse) Oxysterol-binding protein-related protein 11, was expressed in the hosts and tags indicated.
€ 245.00 - € 3405.00
Specifications
Name | Oxysterol-binding protein-related protein 11 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | Osbpl11 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16010971
|
enQuireBio™
QP7816-EC-100UG |
100 μg |
€ 770.00
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16030971
|
enQuireBio™
QP7816-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16020971
|
enQuireBio™
QP7816-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040971
|
enQuireBio™
QP7816-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050971
|
enQuireBio™
QP7816-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060971
|
enQuireBio™
QP7816-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Oxysterol-binding protein-related protein 11 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
Osbpl11 | |
Recombinant Protein | |
E. coli | |
MQGGEPASVM KVSESEGKLE GLATAVTPNK NSGNSSCGGA ISSSSSNSSR GGSAKGWQYS DHMESVNGYL MKYTNLVTGW QYRFFVLNNE AGLLEYFVNE QSRNQKPRGT LQLAGAVISP SDEDSHTFTV NAASGEQYKL RATDAKERQH WVSRLQICTQ HHTEAIGKNN PPLKSRSFSL ASSGNSPISQ RRPSQNAMSF FNVGHSKLQS VNKRAHLHPD HLVEVREMMS HAEGQQRDLI RRIECLPASG LLSSLDQDLL MLKATSMATM NCLNDCFHIL QLQHASHQKG ALPSGTTIEW LEPKIPLSNH YKNGAEQPFA TEPNKPMGAP EAQCVAESGV LAREPEDISA DDEVEDTCDN KEDDLGAVEE QRSVILHLLS QLKLGMDLTR VVLPTFILEK RSLLEMYADF MSHPDLFIGI TNGATPEDRM IRFVEYYLTS FHEGRKGAIA KKPYNPIIGE TFHCSWRMPK SEVASGVSSS SSTPAITDHA PLPEEAPTQS VSDCYTVRFV AEQVSHHPPV SGFYAECAER KMCVNAHVWT KSKFLGMSIG VTMVGEGVLC LLEHGEEYTF SLPCAYARSI LTVPWVELGG KVSVNCAKTG YSASITFHTK PFYGGKLHRV TAEVKYNLTN TVVCRVQGEW NSVLEFTYSN GETKFVDLAK LAVTKKRVRP LEKQDPFESR RLWKNVTDSL RESEIDKATE HKRSLEERQR TEERLRTETG TPWKTKYFIK EGDGWVYHKP LWKGIPSQPA E | |
Greater than 90% as determined by SDS-PAGE. |