missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Oxysterol-binding protein-related protein 11 Protein

A DNA sequence encoding the Mus musculus (Mouse) Oxysterol-binding protein-related protein 11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7816-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030971

  • € 3405.00 / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Oxysterol-binding protein-related protein 11 Protein
Research Use Only
Osbpl11
Recombinant Protein
E. coli
MQGGEPASVM KVSESEGKLE GLATAVTPNK NSGNSSCGGA ISSSSSNSSR GGSAKGWQYS DHMESVNGYL MKYTNLVTGW QYRFFVLNNE AGLLEYFVNE QSRNQKPRGT LQLAGAVISP SDEDSHTFTV NAASGEQYKL RATDAKERQH WVSRLQICTQ HHTEAIGKNN PPLKSRSFSL ASSGNSPISQ RRPSQNAMSF FNVGHSKLQS VNKRAHLHPD HLVEVREMMS HAEGQQRDLI RRIECLPASG LLSSLDQDLL MLKATSMATM NCLNDCFHIL QLQHASHQKG ALPSGTTIEW LEPKIPLSNH YKNGAEQPFA TEPNKPMGAP EAQCVAESGV LAREPEDISA DDEVEDTCDN KEDDLGAVEE QRSVILHLLS QLKLGMDLTR VVLPTFILEK RSLLEMYADF MSHPDLFIGI TNGATPEDRM IRFVEYYLTS FHEGRKGAIA KKPYNPIIGE TFHCSWRMPK SEVASGVSSS SSTPAITDHA PLPEEAPTQS VSDCYTVRFV AEQVSHHPPV SGFYAECAER KMCVNAHVWT KSKFLGMSIG VTMVGEGVLC LLEHGEEYTF SLPCAYARSI LTVPWVELGG KVSVNCAKTG YSASITFHTK PFYGGKLHRV TAEVKYNLTN TVVCRVQGEW NSVLEFTYSN GETKFVDLAK LAVTKKRVRP LEKQDPFESR RLWKNVTDSL RESEIDKATE HKRSLEERQR TEERLRTETG TPWKTKYFIK EGDGWVYHKP LWKGIPSQPA E
Greater than 90% as determined by SDS-PAGE.
1 mg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers