missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse PDAP1 Protein

Product Code. 18122140 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18122140

Brand: Novus Biologicals™ NBP19933110UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results.

  • Recombinant bioactive protein for Mouse PDAP1
  • The Recombinant Mouse PDAP1 Protein is derived from E. coli.
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE
Gene ID (Entrez) 11333
Molecular Weight (g/mol) 28.9 kDa
Purification Method >97%, by SDS-PAGE
Quantity 10 μg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 ng/μg
Gene Symbol PDAP1
Reconstitution Reconstitute with sterile H2O to 0.1 - 0.5 mg/mL, which can be further diluted into other aqueous solutions.
Buffer Lyophilized with no additives.
Purity or Quality Grade >97%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.