missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Protein FAM83D Protein
A DNA sequence encoding the Mus musculus (Mouse) Protein FAM83D, was expressed in the hosts and tags indicated.
€ 287.00 - € 3175.00
Specifications
Accession Number | NP_082251.2 |
---|---|
Gene ID (Entrez) | 71878 |
Name | Protein FAM83D Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028941
|
enQuireBio™
QP9893-YE-100UG |
100 μg |
€ 920.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068941
|
enQuireBio™
QP9893-YE-500UG |
500 μg |
€ 2650.00
500µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038941
|
enQuireBio™
QP9893-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058941
|
enQuireBio™
QP9893-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078941
|
enQuireBio™
QP9893-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048941
|
enQuireBio™
QP9893-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_082251.2 | |
Protein FAM83D Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
71878 | |
Research Use Only | |
Fam83d | |
Recombinant Protein | |
Yeast | |
MAARFELLDD LPAACLSPCG PPNPTELFSE ARRLALEQLL AGGPDAWAAF LRRERLGRFL NADEVREVLG AAERPGEDGA AVAEDSFGSS HECSSGTYFP EQSDLEPPAL ELGWPSFYQG AYRGATRVEA HFQPRGAGAG GPYGCKDALR QQLRSAREVI AVVMDVFSDI DIFRDLQESC RKRGVAVYIL LDQTLLPHFL DMCMDLRVHP EQEKLMTVRT ITGNIYYARS GTKVVGKVHE KFTLIDGIRV ATGSYSFTWT DGKLNSSNLV ILSGQVVEHF DLEFRILYAQ SEPISSKLLS NFQINSKFDH LADRKPQSKE PTLGNLLRMR LARLSSTPRK SNLGPEEPPK DRAKPKRPDS EASTISDEDY FHSHKDQLED SKVADAATQT EPREEMAAVS LSEVGTQTSS SMMCVGTQTT VVTRAASSQA TVWSKSTTTQ TEADESFLPQ GAQSKEGSPA SKMSVSRSSS VRSSSSVSSQ GSLASSVSSH VSLTAADLHT PAYPKYLGLG TPHLDLCLRD SFRNLSKERQ VHFTGIRSRL TQMLTVLSRR TLFTEHYLSY SPGSFTRAST NLVSVRDIAL YPPYQ | |
Greater than 90% as determined by SDS-PAGE. |