missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse R-Spondin 1 (CHO-expressed) Protein

Product Code. 18729846
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18729846

Brand: R&D Systems™ 7150RS010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its ability to induce Topflash reporter activity in HEK293T human embryonic kidney cells. The typical ED50 is 8 to 40 ng/mL in the presence of 5 ng/mL Recombinant Mouse Wnt-3a.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_619624
For Use With (Application) Bioactivity
Gene ID (Entrez) 284654
Molecular Weight (g/mol) 27 kDa
Name Cristin 3, CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPO1, RSPONDIN, RSpondin 1, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse R-Spondin 1 protein Met1-Gln265
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol Rspo1
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 100 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.01 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.