missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SCF / C-kit ligand Protein
Recombinant Protein
€ 2070.00 - € 3330.00
Specifications
Accession Number | NP_001334085.1 |
---|---|
Gene ID (Entrez) | 17311 |
Name | SCF / C-kit ligand Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936069
|
enQuireBio™
QP5410-500UG |
500 μg |
€ 2070.00
500µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926069
|
enQuireBio™
QP5410-1MG |
1 mg |
€ 3330.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001334085.1 | |
SCF / C-kit ligand Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 20 ng/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 5 x 10^4 units/mg. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
17311 | |
Research Use Only | |
Kitl | |
Recombinant Protein | |
E. coli | |
MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA | |
Greater than 95% as determined by SDS-PAGE. |