missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Serine protease inhibitor Kazal-type 1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Serine protease inhibitor Kazal-type 1, was expressed in the hosts and tags indicated.

€ 245.00 - € 3405.00

Specifications

Accession Number NP_033284.1
Gene ID (Entrez) 20730
Name Serine protease inhibitor Kazal-type 1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16088540
View Documents Promotion Details
enQuireBio™
QP7376-EC-100UG
100 μg
€ 770.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16008550
View Documents Promotion Details
enQuireBio™
QP7376-EC-1MG
1 mg
€ 3405.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16098540
View Documents Promotion Details
enQuireBio™
QP7376-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16018550
View Documents Promotion Details
enQuireBio™
QP7376-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16028550
View Documents Promotion Details
enQuireBio™
QP7376-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16038550
View Documents Promotion Details
enQuireBio™
QP7376-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_033284.1
Serine protease inhibitor Kazal-type 1 Protein
Not determined.
Full Length
Mouse
GST
Tris-based buffer, 50% glycerol
20730
Research Use Only
Spink3
Recombinant Protein
E. coli
AKVTGKEASC HDAVAGCPRI YDPVCGTDGI TYANECVLCF ENRKRIEPVL IRKGGPC
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications