missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EpCAM/TROP-1 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB11679-100
This item is not returnable.
View return policy
Description
EpCAM/TROP1 Monoclonal antibody specifically detects EpCAM/TROP1 in Mouse samples. It is validated for Multiplex Immunofluorescence,Immunohistochemistry,Western Blot
Specifications
| EpCAM/TROP1 | |
| Monoclonal | |
| Unconjugated | |
| Q99JW5 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4072 | |
| Mouse | |
| Purified |
| Multiplex Immunofluorescence, Immunohistochemistry, Western Blot | |
| 3153C | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
| Mouse myeloma cell line, NS0-derived mouse EpCAM, Gln24-Thr266, Accession # Q99JW5 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction