missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Thyroid hormone-inducible hepatic Protein
A DNA sequence encoding the Mus musculus (Mouse) Thyroid hormone-inducible hepatic protein, was expressed in the hosts and tags indicated.
€ 245.00 - € 3405.00
Specifications
Accession Number | NP_033407.1 |
---|---|
Gene ID (Entrez) | 21835 |
Name | Thyroid hormone-inducible hepatic Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16000361
|
enQuireBio™
QP7714-EC-100UG |
100 μg |
€ 770.00
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16020361
|
enQuireBio™
QP7714-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010361
|
enQuireBio™
QP7714-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030361
|
enQuireBio™
QP7714-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040361
|
enQuireBio™
QP7714-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050361
|
enQuireBio™
QP7714-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_033407.1 | |
Thyroid hormone-inducible hepatic Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
21835 | |
Research Use Only | |
Thrsp | |
Recombinant Protein | |
E. coli | |
MQVLTKRYPK NCLLTVMDRY SAVVRNMEQV VMIPSLLRDV QLSGPGGSVQ DGAPDLYTYF TMLKSICVEV DHGLLPREEW QAKVAGNETS EAENDAAETE EAEEDRISEE LDLEAQFHLH FCSLHHILTH LTRKAQEVTR KYQEMTGQVL | |
Greater than 90% as determined by SDS-PAGE. |