missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TIE1 Fc Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | TIE1 Fc Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Mouse |
| Species | Mouse |
| Protein Tag | Untagged |
| Sequence | MVWWGSSLLL PTLFLASHVG ASVDLTLLAN LRITDPQRFF LTCVSGEAGA GRSSDPPLLL EKDDRIVRTF PPGQPLYLAR NGSHQVTLRG FSKPSDLVGV FSCVGGAGAR RTRVLYVHNS PGAHLFPDKV THTVNKGDTA VLSAHVHKEK QTDVIWKNNG SYFNTLDWQE ADDGRFQLQL QNVQPPSSGI YSATYLEASP LGSAFFRLIV RGCGAGRWGP GCVKDCPGCL HGGVCHDHDG ECVCPPGFTG TRCEQACREG RFGQSCQEQC PGTAGCRGLT FCLPDPYGCS CGSGWRGSQC QEACAPGHFG ADCRLQCQCQ NGGTCDRFSG CVCPSGWHGV HCEKSDRIPQ ILSMATEVEF NIGTMPRINC AAAGNPFPVR GSMKLRKPDG TMLLSTKVIV EPDRTTAEFE VPSLTLGDSG FWECRVSTSG GQDSRRFKVN VKVPPVPLTA PRLLAKQSRQ LVVSPLVSFS GDGPISSVRL HYRPQDSTIA WSAIVVDPSE NVTLMNLKPK TGYNVRVQLS RPGEGGEGGW GPSALMTTDC PEPLLQPWLE SWHVEGPDRL RVSWSLPSVP LSGDGFLLRL WDGARGQERR ENISFPQART ALLTGLTPGT HYQLDVRLYH CTLLGPASPP AHVHLPPSGP PAPRHLHAQA LSDSEIQLMW QHPEAPSGPI SKYIVEIQVA GGSGDPQWMD VDRPEETSII VRGLNASTRY LFRVRASVQG LGDWSNTVEE ATLGNGLQSE GPVRESRAAE EGLDQQLVLA VVGSVSATCL TILAALLALV CIRRSCLHRR RTFTYQSGSG EETILQFSSG TLTLTRRPKP QPEPLSYPVL EWEDITFEDL IGEGNFGQVI RAMIKKDGLK MNAAIKMLKE YASENDHRDF AGELEVLCKL GHHPNIINLL GACENRGYLY IAIEYAPYGN LLDFLRKSRV LETDPAFARE HGTASTLSSR QLLRFASDAA NGMQYLSEKQ FIHRDLAARN VLVGENLASK IADFGLSRGE EVYVKKTMGR LPVRWMAIES LNYSVYTTKS DVWSFGVLLW EIVSLGGTPY CGMTCAELYE KLPQGYRMEQ PRNCDDEVYE LMRQCWRDRP YERPPFAQIA LQLGRMLEAR KAYVNMSLFE NFTYAGIDAT AEEA |
| Buffer | TIE-1 Fc Chimera was lyophilized from a concentrated (1 mg/ml) sterile solution containing 1x PBS. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction