missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TMEFF2/Tomoregulin-2 Fc Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Mouse PDGF-AB (Catalog # 10839-AB ) is immobilized at 2 μg/mL (100 μL/well), Recombinant Mouse TMEFF2/Tomoregulin-2 Fc Chimera (Catalog # 11204-TF) binds with an ED50 of 1.00-9.00 μg/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 23671 |
| Molecular Weight (g/mol) | M.W.-observed: 77-90 kDa, under reducing conditions., M.W.-theroretical: 58 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse TMEFF2/Tomoregulin-2 protein Mouse TMEFF2 (Phe41-Val320) (C-terminus) Accession # Q9QYM9.1 IEGRMDP Mouse IgG2A (Glu98-Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CT120.2, HPP1member 2, Hyperplastic polyposis protein 1, TPEFtomoregulin, TR-2, transmembrane protein TENB2, Transmembrane protein with EGF-like and two follistatin-like domains, transmembrane protein with EGF-like and two follistatin-like domains 2, TRtomoregulin-2 |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction