missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TNFRSF4 / OX40 / CD134 Protein

Product Code. 16076040
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16076040 100 μg 100µg
16086040 10 μg 10µg
16026050 50 μg 50µg
16006050 200 μg 200µg
16016050 500 μg 500µg
16096040 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16076040 Supplier enQuireBio™ Supplier No. QP6816ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) TNFRSF4 / OX40 / CD134, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_035789.1
Gene ID (Entrez) 22163
Name TNFRSF4 / OX40 / CD134 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TNFRSF4
Protein Length Extracellular Domain
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His-SUMO
Sequence VTARRLNCVK HTYPSGHKCC RECQPGHGMV SRCDHTRDTL CHPCETGFYN EAVNYDTCKQ CTQCNHRSGS ELKQNCTPTQ DTVCRCRPGT QPRQDSGYKL GVDCVPCPPG HFSPGNNQAC KPWTNCTLSG KQTRHPASDS LDAVCEDRSL LATLLWETQR PTFRPTTVQS TTVWPRTSEL PSPPTLVTPE GP
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.