missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse UCHL3 Protein
A cDNA sequence encoding the UCHL3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10905-2ug
Additional Details : Weight : 0.01000kg
Specifications
50933 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 9,000 pmol/min/mg, and is defined as the amount of enzyme that hydrolysis 1.0 pmole of ubiquitin-AMC per minute at pH 7.5, at 37C. | |
Mouse | |
His | |
UCHL3 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol. |
UCHL3 Protein | |
Research Use Only | |
Uchl3 | |
Recombinant Protein | |
Baculovirus | |
MEGQRWLPLE ANPEVTNQFL KQLGLHPNWQ FVDVYGMEPE LLSMVPRPVC AVLLLFPITE KYEVFRTEEE EKIKSQGQDV TSSVYFMKQT ISNACGTIGL IHAIANNKDK MHFESGSTLK KFLEESVSMS PEERAKFLEN YDAIRVTHET SAHEGQTEAP SIDEKVDLHF IALVHVDGHL YELDGRKPFP INHGKTSDET LLEDAIEVCK KFMERDPDEL RFNAIALSAA LEHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |