missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse UDP glucose dehydrogenase Protein

Product Code. 18211302 Shop All Bio Techne Products
Click to view available options
Quantity:
20 μg
5 μg
Unit Size:
20µg
5µg
This item is not returnable. View return policy

Product Code. 18211302

Brand: Novus Biologicals™ NBP2614525UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a N-terminalHis-tag and corresponding to the amino acids 1-493 of Mouse UDP glucose dehydrogenase. The Recombinant Mouse UDP glucose dehydrogenase Protein is derived from E. coli. The Recombinant Mouse UDP glucose dehydrogenase Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation 20mM MES buffer (pH5.0) containing 20% glycerol, 150mM NaCl, 1mM EDTA
Gene ID (Entrez) 7358
Molecular Weight (g/mol) 57.2 kDa
Quantity 5 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1μg of protein (determined by LAL method)
Gene Symbol μgDH
Cross Reactivity UDP glucose dehydrogenase
Species Mouse
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.