missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Vomeronasal secretory protein 1

A DNA sequence encoding the Mus musculus (Mouse) Vomeronasal secretory protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7718-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16000391

  • € 985.00 / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_034824.1
Vomeronasal secretory protein 1
Research Use Only
Lcn3
Recombinant Protein
E. coli
QDSSFLAFNN GNFSGKWFLK ALVSEDDIPI NKVSPMLILV LNNGDIELSI THMIYDQCLE VTTILEKTDV PGQYLAFEGK THLQVQLSSV KGHYMLYCDG EIEGMRFLMT QLIGRDPQEN LEALEEFKVF TQIKGLVAEN LVILEQMEKC EPESFYELPS RPSE
Greater than 90% as determined by SDS-PAGE.
16820
100 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers