missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Vomeronasal secretory protein 1

A DNA sequence encoding the Mus musculus (Mouse) Vomeronasal secretory protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7718-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040391

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_034824.1
Vomeronasal secretory protein 1
Research Use Only
Lcn3
Recombinant Protein
E. coli
QDSSFLAFNN GNFSGKWFLK ALVSEDDIPI NKVSPMLILV LNNGDIELSI THMIYDQCLE VTTILEKTDV PGQYLAFEGK THLQVQLSSV KGHYMLYCDG EIEGMRFLMT QLIGRDPQEN LEALEEFKVF TQIKGLVAEN LVILEQMEKC EPESFYELPS RPSE
Greater than 90% as determined by SDS-PAGE.
16820
500 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers