Learn More
enQuireBio™ Recombinant Multi Species Buserelin Protein
Recombinant Protein
Brand:  enQuireBio™ QP11214-5mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15946508
  • € 402.00 / 5mg
Estimated Shipment: 28-06-2024
to see stock.
Specifications
Multi Species Buserelin Protein
Research Use Only
Recombinant Protein
Untagged
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
5 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Multi Species
The Buserelin peptide was lyophilized with no additives.