Learn More
enQuireBio™ Recombinant Multi Species Buserelin Protein
Recombinant Protein
Brand:  enQuireBio™ QP11214-20mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15936508
  • € 1040.00 / 20mg
Estimated Shipment: 06-02-2023
to see stock.
Add to basket
Specifications
Multi Species Buserelin Protein
20 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Multi Species
The Buserelin peptide was lyophilized with no additives.
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
Untagged
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers