missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Aminopeptidase Protein
Click to view available options
Quantity:
100 IU
250 U
500 U
Unit Size:
100 units
250 units
500 units
Specifications
Specifications
| Name | Aminopeptidase Protein |
| Quantity | 250 U |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | Recombinant Aeromonas Aminopeptidase was found to have an activity of 120 Units/mg protein. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | Non-species Specific |
| Protein Tag | Untagged |
| Sequence | MKYTKTLLAM VLSATFCQAY AEDKVWISIG ADANQTVMKS GAESILPNSV ASSGQVWVGQ VDVAQLAELS HNMHEEHNRC GGYMVHPSAQ SAMAASAMPT TLASFVMPPI TQQATVTAWL PQVDASQITG TISSLESFTN RFYTTTSGAQ ASDWIASEWQ ALSASLPNAS VKQVSHSGYN QKSVVMTITG SEAPDEWIVI GGHLDSTIGS HTNEQSVAPG ADDDASGIAA VTEVIRVLSE NNFQPKRSIA FMAYAAEEVG LRGSQDLANQ YKSEGKNVVS ALQLDMTNYK GSAQDVVFIT DYTDSNFTQY LTQLMDEYLP SLTYGFDTCG YACSDHASWH NAGYPAAMPF ESKFNDYNPR IHTTQDTLAN SDPTGSHAKK FTQLGLAYAI EMGSATGDTP TPGNQLEDGV PVTDLSGSRG SNVWYTFELE TQKNLQITTS GGYGDLDLYV KFGSKASKQN WDCRPYLSGN NEVCTFNNAS PGTYSVMLTG YSNYSGASLK ASTF |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction