missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Atosiban Protein
A cDNA sequence encoding the Atosiban was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11102-1gram
Additional Details : Weight : 0.01000kg
Specifications
Atosiban Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Atosiban peptide was lyophilized with no additives. |
1 g | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 99.0% as determined by analysis by RP-HPLC. |