missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Atosiban Protein
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
Specifications
Specifications
| Name | Atosiban Protein |
| Quantity | 50 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | Untagged |
| Buffer | The Atosiban peptide was lyophilized with no additives. |
| Purity or Quality Grade | Greater than 99.0% as determined by analysis by RP-HPLC. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction