Learn More
enQuireBio™ Recombinant other Avidin Biotin Protein
Product Code. 15984138
Click to view available options
Quantity:
2 mg
10 mg
100 mg
Unit Size:
100mg
10mg
2mg
Product Code. 15984138
Brand: enQuireBio™ QP105262mg
Recombinant Protein
Specifications
Gene ID (Entrez) 396260
Name Avidin Biotin Protein
Quantity 2 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol AVD
Biological Activity Binding activity of Avidin-Biotin is 10-15units/mg protein. One unit of Avidin activity is defined as the amount of protein which will bind 1microg d-biotin.
Product Type Recombinant Protein
Cross Reactivity Other
Protein Tag Untagged
Sequence MVHATSPLLL LLLLSLALVA PGLSARKCSL TGKWTNDLGS NMTIGAVNSR GEFTGTYITA VTATSNEIKE SPLHGTQNTI NKRTQPTFGF TVNWKFSEST TVFTGQCFID RNGKEVLKTM WLLRSSVNDI GDDWKATRVG INIFTRLRTQ KE
Buffer Salt-free lyophilized powder with no preservative added.