missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Avidin Biotin Protein
Click to view available options
Quantity:
2 mg
10 mg
100 mg
Unit Size:
100mg
10mg
2mg
Specifications
Specifications
| Gene ID (Entrez) | 396260 |
| Name | Avidin Biotin Protein |
| Quantity | 10 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Gene Symbol | AVD |
| Biological Activity | Binding activity of Avidin-Biotin is 10-15units/mg protein. One unit of Avidin activity is defined as the amount of protein which will bind 1microg d-biotin. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Protein Tag | Untagged |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction