missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Borrelia Afzelii DbpA Protein
A cDNA sequence encoding the Borrelia Afzelii DbpA was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 5450.00
Specifications
Name | Borrelia Afzelii DbpA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946388
|
enQuireBio™
QP11177-5UG |
5 μg |
€ 137.00
5µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936388
|
enQuireBio™
QP11177-20UG |
20 μg |
€ 212.00
20µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926388
|
enQuireBio™
QP11177-1MG |
1 mg |
€ 5450.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Borrelia Afzelii DbpA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 80.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Borrelia Afzelii DbpA is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |