missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Car b 1.0109 Protein
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
Specifications
Specifications
| Name | Car b 1.0109 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGVFNYEAET PSVIPAARLF KSYVLDGDKL IPKVAPQAIS SVENVGGNGG PGTIKNITFA EGSPFKFVKE RVDEVDNANF KYNYTVIEGD VLGDKLEKVS HELKIVAAPG GGSILKISSK FHAKGDHEVN AEEMKGAKEM AEKLLRAVES YLLAHTAEYN |
| Buffer | Car b 1.0109 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction