missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Cetrorelix Protein
Click to view available options
Quantity:
1 mg
10 mg
250 μg
Unit Size:
10mg
1mg
250µg
Specifications
Specifications
| Name | Cetrorelix Protein |
| Quantity | 250 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | Untagged |
| Buffer | The Cetrorelix peptide was lyophilized with no additives. |
| Purity or Quality Grade | Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction