missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Cetrorelix Protein
A cDNA sequence encoding the Cetrorelix was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 1210.00
Specifications
Name | Cetrorelix Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15957128
|
enQuireBio™
QP11394-250UG |
250 μg |
€ 137.00
250µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947128
|
enQuireBio™
QP11394-1MG |
1 mg |
€ 212.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937128
|
enQuireBio™
QP11394-10MG |
10 mg |
€ 1210.00
10mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Cetrorelix Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Cetrorelix peptide was lyophilized with no additives. |