missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Cyclophilin A Protein

Product Code. 15997688
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15997688

Brand: enQuireBio™ QP1157620ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Cyclophilin A was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 947870
Name Cyclophilin A Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol ppiA
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSAKGDPHV LLTTSAGNIE LELDKQKAPV SVQNFVDYVN SGFYNNTTFH RVIPGFMIQG GGFTEQMQQK KPNPPIKNEA DNGLRNTRGT IAMARTADKD SATSQFFINV ADNAFLDHGQ RDFGYAVFGK VVKGMDVADK ISQVPTHDVG PYQNVPSKPV VILSAKVLP
Buffer Cyclophilin A protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.