missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Der F1 Protein
A cDNA sequence encoding the Der F1 was constructed and used to recombinantly synthesize the protein.
€ 238.00 - € 1815.00
Specifications
Name | Der F1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | DERF1 |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15917938
|
enQuireBio™
QP11656-100UG |
100 μg |
€ 238.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937938
|
enQuireBio™
QP11656-500UG |
500 μg |
€ 910.00
500µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927938
|
enQuireBio™
QP11656-1MG |
1 mg |
€ 1815.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Der F1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKFVLAIASL LVLSTVYARP ASIKTFEEFK KAFNKNYATV EEEEVARKNF LESLKYVEAN KGAINHLSDL SLDEFKNRYL MSAEAFEQLK TQFDLNAETS ACRINSVNVP SELDLRSLRT VTPIRMQGGC GSCWAFSGVA ATESAYLAYR NTSLDLSEQE LVDCASQHGC HGDTIPRGIE YIQQNGVVEE RSYPYVAREQ RCRRPNSQHY GISNYCQIYP PDVKQIREAL TQTHTAIAVI IGIKDLRAFQ HYDGRTIIQH DNGYQPNYHA VNIVGYGSTQ GDDYWIVRNS WDTTWGDSGY GYFQAGNNLM MIEQYPYVVI M | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |
Research Use Only | |
DERF1 | |
Other | |
His | |
60mM NaCl, 50mM Tris-HCl pH 8.0 and 1.2M Urea. |