missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DnaJ Protein
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
Specifications
Specifications
| Name | DnaJ Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MAKQDYYEIL GVSKTAEEHE IRKAYKRLAM KYHPDRNQGD KEAEAKFKEI KEAYEVLTDSQKRAAYDQYG HAAFEQGGMG GGGFGGGADF SDIFGDVFGD IFGGGRGRQR AARGADLRYNMELTLEEAVR GVTKEIRIPT LEECDVCHGS GAKPGTQPQT CPTCHGSGQV QMRQGFFAVQQTCPHCQGRG TLIKDPCNKC HGHGRVERSK TLSVKIPAGV DTGDRIRLAG EGEAGEHGAPAGDLYVQVQV KQHPIFEREG NNLYCEVPIN FAMAALGGEI EVPTLDGRVK LKVPGETQTG KLFRMRGKGV KSVRGGAQGD LLCRVVVETP VGLNERQKQL LQELQESFGG PTGEHNSPRSKSFFDGVKKF FDDLTR |
| Buffer | The DnaJ contains 25mM Tris-HCl buffer (pH 7.5), 100mM NaCl, 5mM DTT and 10% Glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction