Learn More
enQuireBio™ Recombinant other Gly m 4.0101 Protein
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
Specifications
Specifications
| Name | Gly m 4.0101 Protein |
| Quantity | 50 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | Baculovirus |
| Protein Tag | His |
| Sequence | MGVFTFEDEI NSPVAPATLY KALVTDADNV IPKALDSFKS VENVEGNGGP GTIKKITFLE DGETKFVLHK IESIDEANLG YSYSVVGGAA LPDTAEKITF DSKLVAGPNG GSAGKLTVKY ETKGDAEPNQ DELKTGKAKA DALFKAIEAY LLAHPDYN |
| Buffer | Gly m 4.0101 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
| Show More |