Learn More
enQuireBio™ Recombinant other Gly m 5.0101 Protein
A cDNA sequence encoding the Gly m 5.0101 was constructed and used to recombinantly synthesize the protein.
€ 910.00 - € 6055.00
Specifications
Name | Gly m 5.0101 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15949048
|
enQuireBio™
QP11998-50UG |
50 μg |
€ 910.00
50µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929048
|
enQuireBio™
QP11998-100UG |
100 μg |
€ 1365.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939048
|
enQuireBio™
QP11998-1MG |
1 mg |
€ 6055.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Gly m 5.0101 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Gly m 5.0101 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
VEKEECEEGE IPRPRPRPQH PEREPQQPGE KEEDEDEQPR PIPFPRPQPR QEEEHEQREE QEWPRKEEKR GEKGSEEEDE DEDEEQDERQ FPFPRPPHQK EERKQEEDED EEQQRESEES EDSELRRHKN KNPFLFGSNR FETLFKNQYG RIRVLQRFNQ RSPQLQNLRD YRILEFNSKP NTLLLPNHAD ADYLIVILNG TAILSLVNND DRDSYRLQSG DALRVPSGTT YYVVNPDNNE NLRLITLAIP VNKPGRFESF FLSSTEAQQS YLQGFSRNIL EASYDTKFEE INKVLFSREE GQQQGEQRLQ ESVIVEISKE QIRALSKRAK SSSRKTISSE DKPFNLRSRD PIYSNKLGKF FEITPEKNPQ LRDLDIFLSI VDMNEGALLL PHFNSKAIVI LVINEGDANI ELVGLKEQQQ EQQQEEQPLE VRKYRAELSE QDIFVIPAGY PVVVNATSNL NFFAIGINAE NNQRNFLAGS QDNVISQIPS QVQELAFPGS AQAVEKLLKN QRESYFVDAQ PKKKEEGNKG RKGPLSSILR AFY | |
Greater than 80.0% as determined by SDS-PAGE. |