Learn More
enQuireBio™ Recombinant other HPSE Active Protein
Product Code. 15944588
Click to view available options
Quantity:
1 μg
16 μg
4 μg
Unit Size:
16µg
1µg
4µg
Product Code. 15944588
Brand: enQuireBio™ QP1066816ug
Recombinant Protein
Specifications
Name HPSE Active Protein
Quantity 16 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The specific activity of Heparanase Active Enzyme in-house standard is about 0.7 Units (1 unit = 1 umole of reducing ends of heparan sulfate substrate formed per minute per mg Heparanase Active Enzyme at 37C). The enzymatic activity of each Heparanase Active Enzyme batch is comparable to the standard as determined by activity assay in which immobilized heparan, released due to heparanase activity, is quantified colorimetrically. Recommended reaction buffer: 20 mM Citrate Phosphate buffer, pH 5.4; 50mM NaCl; 1mM CaCl2.
Product Type Recombinant Protein
Cross Reactivity Other
Protein Tag Untagged
Buffer rate Phosphate buffer, pH 5.4; 50mM NaCl; 1mM CaCl2.
Purity or Quality Grade Greater than 95.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.