missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other HPSE WB Protein

Product Code. 15939998
Click to view available options
Quantity:
100 ng
200 ng
300 ng
Unit Size:
0.1µg
0.2µg
This item is not returnable. View return policy

Product Code. 15939998

Brand: enQuireBio™ QP12305100ng

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Gene ID (Entrez) 10855
Name HPSE WB Protein
Quantity 100 ng
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol HPSE
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag Untagged
Sequence MLLRSKPALP PPLMLLLLGP LGPLSPGALP RPAQAQDVVD LDFFTQEPLH LVSPSFLSVT IDANLATDPR FLILLGSPKL RTLARGLSPA YLRFGGTKTD FLIFDPKKES TFEERSYWQS QVNQDICKYG SIPPDVEEKL RLEWPYQEQL LLREHYQKKF KNSTYSRSSV DVLYTFANCS GLDLIFGLNA LLRTADLQWN SSNAQLLLDY CSSKGYNISW ELGNEPNSFL KKADIFINGS QLGEDFIQLH KLLRKSTFKN AKLYGPDVGQ PRRKTAKMLK SFLKAGGEVI DSVTWHHYYL NGRTATKEDF LNPDVLDIFI SSVQKVFQVV ESTRPGKKVW LGETSSAYGG GAPLLSDTFA AGFMWLDKLG LSARMGIEVV MRQVFFGAGN YHLVDENFDP LPDYWLSLLF KKLVGTKVLM ASVQGSKRRK LRVYLHCTNT DNPRYKEGDL TLYAINLHNV TKYLRLPYPF SNKQVDKYLL RPLGPHGLLS KSVQLNGLTL KMVDDQTLPP LMEKPLRPGS SLGLPAFSYS FFVIRNAKVA ACI
Buffer Concentration: 1ug /ml Content: 100 ngBuffer: LDS-PAGE buffer[140 mM Tris buffer pH 8.5, 10% Glycerol, 2% LDS, 0.015% EDTA, 1.88% (v/v) of 1% Serva Blue G250 and 0.625% (v/v) of 1% Phenol red].
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.