missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other LLO Protein

Product Code. 15985118
Click to view available options
Quantity:
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
50µg
This item is not returnable. View return policy

Product Code. 15985118

Brand: enQuireBio™ QP1077310ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the LLO was constructed and used to recombinantly synthesize the protein.

Specifications

Name LLO Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol hlyA
Biological Activity Hemolytic activity for Recombinant Listeriolysin O was 1.18 x 105 HU/mg of protein where HU means hemolytic activity unit that is the amount of toxin needed to release half the hemoglobin (50% lysis) of the erythrocytes as determined by hemolysin assay. 2mM DTT is used to reactivate the toxin.
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag Untagged
Sequence MKKIMLVFIT LILISLPIAQ QTEAKDASAF HKEDLISSMA PPTSPPASPK TPIEKKHADE IDKYIQGLDY NKNNVLVYHG DAVTNVPPRK GYKDGNEYIV VEKKKKSINQ NNADIQVVNA ISSLTYPGAL VKANSELVEN QPDVLPVKRD SLTLSIDLPG MTNQDNKIVV KNATKSNVNN AVNTLVERWN EKYAQAYPNV SAKIDYDDEM AYSESQLIAK FGTAFKAVNN SLNVNFGAIS EGKMQEEVIS FKQIYYNVNV NEPTRPSRFF GKAVTKEQLQ ALGVNAENPP AYISSVAYGR QVYLKLSTNS HSTKVKAAFD AAVSGKSVSG DVELTNIIKN SSFKAVIYGG SAKDEVQIID GNLGDLRDIL KKGATFNRET PGVPIAYTTN FLKDNELAVI KNNSEYIETT SKAYTDGKIN IDHSGGYVAQ FNISWDEINY DPEGNEIVQH KNWSENNKSK LAHFTSSIYL PGNARNINVY AKECTGLAWE WWRTVIDDRN LPLVKNRNIS IWGTTLYPKY SNSVDNPIE
Buffer The protein contains 50mM NaH2PO4, 1mM EDTA, 2.7mM KCl, 1mM DTT, 5% (v/v) glycerol and 0.5M NaCl.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.