missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Lysostaphin Protein
A cDNA sequence encoding the Lysostaphin was constructed and used to recombinantly synthesize the protein.
€ 136.00 - € 570.00
Specifications
Name | Lysostaphin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | lss |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935138
|
enQuireBio™
QP10778-1MG |
1 mg |
€ 136.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945138
|
enQuireBio™
QP10778-5MG |
5 mg |
€ 349.00
5mg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925138
|
enQuireBio™
QP10778-10MG |
10 mg |
€ 570.00
10mg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Lysostaphin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MKKTKNNYYT RPLAIGLSTF ALASIVYGGI QNETHASEKS NMDVSKKVAE VETSKAPVEN TAEVETSKAP VENTAEVETS KAPVENTAEV ETSKAPVENT AEVETSKAPV ENTAEVETSK APVENTAEVE TSKAPVENTA EVETSKAPVE NTAEVETSKA PVENTAEVET SKAPVENTAE VETSKAPVEN TAEVETSKAP VENTAEVETS KAPVENTAEV ETSKAPVENT AEVETSKALV QNRTALRAAT HEHSAQWLNN YKKGYGYGPY PLGINGGMHY GVDFFMNIGT PVKAISSGKI VEAGWSNYGG GNQIGLIEND GVHRQWYMHL SKYNVKVGDY VKAGQIIGWS GSTGYSTAPH LHFQRMVNSF SNSTAQDPMP FLKSAGYGKA GGTVTPTPNT GWKTNKYGTL YKSESASFTP NTDIITRTTG PFRSMPQSGV LKAGQTIHYD EVMKQDGHVW VGYTGNSGQR IYLPVRTWNK STNTLGVLWG TIK | |
97% as determined by RP-HPLC. |
Research Use Only | |
lss | |
Other | |
Untagged | |
The protein was lyophilized without any additives. |