Learn More
enQuireBio™ Recombinant other MT I Protein
Product Code. 15911409
Click to view available options
Quantity:
20 mg
100 mg
500 mg
Unit Size:
100mg
20mg
500mg
Product Code. 15911409
Brand: enQuireBio™ QP1274520mg
Recombinant Protein
Specifications
Name MT I Protein
Quantity 20 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag Untagged
Buffer The protein (1 mg/ml) was lyophilized with no additives.
Purity or Quality Grade Greater than 98.0% as determined by RP-HPLC.