missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other PCC Protein

A cDNA sequence encoding the PCC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12973-300IU

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942119

  • € 930.00 / 300 units

Estimated Shipment: 30-01-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PCC Protein
Research Use Only
Recombinant Protein
E. coli
Lyophilized Powder from 0.1M sodium chloride and 10mM sodium citrate buffer.
300 IU
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Videos
SDS
Documents
Certificates
Special Offers

Special Offers