missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other PhI p 2 Protein

A cDNA sequence encoding the PhI p 2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13041-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932329

  • € 800.00 / 50µg

Estimated Shipment: 03-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PhI p 2 Protein
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
PhI p 2 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
MSMASSSSSS LLAMAVLAAL FAGAWCVPKV TFTVEKGSNE KHLAVLVKYE GDTMAEVELR EHGSDEWVAM TKGEGGVWTF DSEEPLQGPF NFRFLTEKGM KNVFDDVVPE KYTIGATYAP EE
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers