missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other PhI p 5 Protein

Product Code. 15952329
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
This item is not returnable. View return policy

Product Code. 15952329

Brand: enQuireBio™ QP130421mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PhI p 5 was constructed and used to recombinantly synthesize the protein.

Specifications

Name PhI p 5 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
Species Baculovirus
Protein Tag His
Sequence SVKRSNGSAE VHRGAVPRRG PRGGPGRSYA ADAGYAPATP AAAGAEAGKA TTEEQKLIED INVGFKAAVA AAASVPAADK FKTFEAAFTS SSKAATAKAP GLVPKLDAAY SVAYKAAVGA TPEAKFDSFV ASLTEALRVI AGALEVHAVK PVTEEPGMAK IPAGELQIID KIDAAFKVAA TAAATAPADD KFTVFEAAFN KAIKESTGGA YDTYKCIPSL EAAVKQAYAA TVAAAPQVKY AVFEAALTKA ITAMSEVQKV SQPATGAATV AAGAATTAAG AASGAATVAA GGYKV
Buffer PhI p 5 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Purity or Quality Grade Greater than 80.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.