missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other PROK Tritirachium album Protein
Click to view available options
Quantity:
20 mg
100 mg
1 g
Unit Size:
100mg
1g
20mg
Specifications
Specifications
| Name | PROK Tritirachium album Protein |
| Quantity | 100 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Gene Symbol | PROK |
| Biological Activity | 34 Units/mg protein. One unit is defined as the amount of enzyme that will hydrolyze urea-denatured hemoglobin to produce color equivalent to 1.0 mol tyrosine per min at 37C, pH 7.5 (color by Folin-Ciocalteu reagent). |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | Yeast |
| Protein Tag | Untagged |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction