missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Staphylokinase Protein
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
Specifications
Specifications
| Name | Staphylokinase Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | The biological activity measured by the ability of fibrin lysis in agarose plate was found to be 50,000IU/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MLKRSLLFLT VLLLLFSFSS ITNEVSASSS FDKGKYKKGD DASYFEPTGP YLMVNVTGVD GKGNELLSPH YVEFPIKPGT TLTKEKIEYY VEWALDATAY KEFRVVELDP SAKIEVTYYD KNKKKEETKS FPITEKGFVV PDLSEHIKNP GFNLITKVVI EKK |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction